Anti-DFFA Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13556
10710-586EA 1178.46 USD
10710-586
Anti-DFFA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Dffa.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: DFFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR