Anti-CCDC86 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23751
10706-126EA 428.55 USD
10706-126
Anti-CCDC86 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CCDC86.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: CCDC86
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR