Anti-Streptococcus agalactiae Mouse Monoclonal Antibody [clone: BDI560]

Supplier: Abnova
MAB4341
10628-816EA 825 USD
10628-816
Anti-Streptococcus agalactiae Mouse Monoclonal Antibody [clone: BDI560]
Antibodies
Mouse monoclonal antibody raised against Streptococcus agalactiae.

Type: Primary
Antigen: Streptococcus agalactiae
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: BDI560
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Streptococcus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR