Anti-PPP1R3D Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23755
10729-924EA 527.69 USD
10729-924
Anti-PPP1R3D Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant PPP1R3D.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: PPP1R3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR