Anti-MME Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: LT10]

Supplier: Abnova
MAB4348
10725-598EA 329 USD
10725-598
Anti-MME Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: LT10]
Antibodies
Mouse monoclonal antibody raised against native Mme.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: LT10
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR