Anti-ITGAX Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BU15]

Supplier: Abnova
MAB4350
10719-774EA 406.15 USD
10719-774
Anti-ITGAX Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BU15]
Antibodies
Mouse monoclonal antibody raised against native ITGAX.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: BU15
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR