Anti-BIRC5 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13568
10691-294EA 1178.46 USD
10691-294
Anti-BIRC5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Birc5.

Recommended Dilutions: Western Blot: 2.5-5 ?g/ml; Immunohistochemistry: 10 ?g/ml

Type: Primary
Antigen: BIRC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR