Anti-HSP90AA1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13569
10735-138EA 1572.31 USD
10735-138
Anti-HSP90AA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Hsp90aa1.

Recommended Dilutions: Western Blot: 5-10 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5-10 ?g/ml; Immunoprecipitation: 10 ?g/mg protein lysate)

Type: Primary
Antigen: HSP90AA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR