Anti-WHSC1L1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11983
10744-600EA 786.15 USD
10744-600
Anti-WHSC1L1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of WHSC1L1.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: WHSC1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR