Anti-UCHL1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11987
10743-732EA 653.85 USD
10743-732
Anti-UCHL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of UCHL1.

Recommended Dilutions: Western Blot: 0.5-2 ?g/ml; Immunohistochemistry: 2.5-5 ?g/ml

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Guinea Pig, Horse, Human, Mouse, Pig, Primates, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR