Anti-TUBB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11989
10743-312EA 653.85 USD
10743-312
Anti-TUBB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TUBB.

Type: Primary
Antigen: TUBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse, Pig, Primates, Rat, Xenopus, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR