Anti-TRPM2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11991
10742-978EA 530.32 USD
10742-978
Anti-TRPM2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Trpm2.

Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: TRPM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR