Anti-Tankyrase Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11996
10742-262EA 319 USD
10742-262
Anti-Tankyrase Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TNKS.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: Tankyrase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR