Anti-TMED10 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11998
10741-664EA 530.32 USD
10741-664
Anti-TMED10 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TMED10.

Recommended Dilutions: Western Blot: 2 ?g/ml; Immunohistochemistry: 1 ?g/ml

Type: Primary
Antigen: TMED10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Primates, Rabbit, Rat, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR