Anti-TLR6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11999
10741-560EA 319 USD
10741-560
Anti-TLR6 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of TLR6.

Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: TLR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Primates, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR