Anti-TLR10 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12001
10741-398EA 415 USD
10741-398
Anti-TLR10 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of TLR10.

Recommended Dilutions: Western Blot: 2 mg/ml

Type: Primary
Antigen: TLR10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR