Anti-STRN Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12016
10738-222EA 653.85 USD
10738-222
Anti-STRN Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of STRN.

Recommended Dilutions: Immunofluorescence: 1:5000; Western Blot: 1:500-1:5000

Type: Primary
Antigen: STRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR