Anti-SSTR1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12019
10739-548EA 653.85 USD
10739-548
Anti-SSTR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Sstr1.

Recommended Dilutions: Western Blot: 1:1000-1:5000

Type: Primary
Antigen: SSTR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR