Anti-CASP2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB1202
10705-642EA 406.15 USD
10705-642
Anti-CASP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CASP2.

Recommended Dilutions: Immunoprecipitation: 1 ?l

Type: Primary
Antigen: CASP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR