Anti-Corynebacterium diphtheriae Diphtheria Toxin (A subunit) Mouse Monoclonal Antibody [clone: 8A4]

Supplier: Abnova
MAB2824
10676-176EA 319 USD
10676-176
Anti-Corynebacterium diphtheriae Diphtheria Toxin (A subunit) Mouse Monoclonal Antibody [clone: 8A4]
Antibodies
Mouse monoclonal antibody raised against Corynebacterium diphtheriae Diphtheria Toxin (A subunit).

Type: Primary
Antigen: Corynebacterium diphtheriae Diphtheria Toxin (A subunit)
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 8A4
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR