Anti-GHRL Monoclonal Antibody [clone: Ghr5]

Supplier: Abnova
MAB2827
10681-884EA 515 USD
10681-884
Anti-GHRL Monoclonal Antibody [clone: Ghr5]
Antibodies
Mouse monoclonal antibody raised against Ghrelin.

Type: Primary
Antigen: Ghrelin
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: Ghr5
Epitope:
Host: IgG2a
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR