Anti-HCG Monoclonal Antibody [clone: G2]

Supplier: Abnova
MAB2831
10676-412EA 427.45 USD
10676-412
Anti-HCG Monoclonal Antibody [clone: G2]
Antibodies
Mouse monoclonal antibody raised against human chorionic gonadotropin (hCG).

Type: Primary
Antigen: chorionic gonadotropin
Clonality: Monoclonal
Clone: G2
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR