Anti-HCG Monoclonal Antibody [clone: G9]

Supplier: Abnova
MAB2832
10676-414EA 319 USD
10676-414
Anti-HCG Monoclonal Antibody [clone: G9]
Antibodies
Mouse monoclonal antibody raised against human chorionic gonadotropin (hCG).

Type: Primary
Antigen: chorionic gonadotropin
Clonality: Monoclonal
Clone: G9
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR