Anti-IgG Rabbit Monoclonal Antibody (Biotin)

Supplier: Abnova
PAB10884
10735-984EA 426.15 USD
10735-984
Anti-IgG Rabbit Monoclonal Antibody (Biotin)
Antibodies
Rabbit anti-rat IgG recognizes rat IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope: F(c)
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR