Anti-HCG Monoclonal Antibody [clone: 77F12]

Supplier: Abnova
MAB2835
10676-408EA 527.69 USD
10676-408
Anti-HCG Monoclonal Antibody [clone: 77F12]
Antibodies
Mouse monoclonal antibody raised against human chorionic gonadotropin (hCG).

Type: Primary
Antigen: chorionic gonadotropin
Clonality: Monoclonal
Clone: 77F12
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR