Anti-Rat IgG Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Abnova
PAB10886
10638-114EA 216.25 USD
10638-114
Anti-Rat IgG Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Sheep anti-rat IgG recognizes rat IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Rhodamine.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host:
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR