Anti-Insulin/Proinsulin Mouse Monoclonal Antibody [clone: D3E7]

Supplier: Abnova
MAB2845
10676-690EA 406.15 USD
10676-690
Anti-Insulin/Proinsulin Mouse Monoclonal Antibody [clone: D3E7]
Antibodies
Mouse monoclonal antibody raised against Insulin/Proinsulin.

Type: Primary
Antigen: Insulin/Proinsulin
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: D3E7
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity: Bovine, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR