Anti-P4 Monoclonal Antibody [clone: XM207]

Supplier: Abnova
MAB2858
10677-100EA 406.15 USD
10677-100
Anti-P4 Monoclonal Antibody [clone: XM207]
Antibodies
Mouse monoclonal antibody raised against Progesterone.

Type: Primary
Antigen: P4
Clonality: Monoclonal
Clone: XM207
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR