Anti-BSG Mouse Monoclonal Antibody [clone: MEM-M6/2]

Supplier: Abnova
MAB3633
10691-580EA 329 USD
10691-580
Anti-BSG Mouse Monoclonal Antibody [clone: MEM-M6/2]
Antibodies
Mouse monoclonal antibody raised against recombinant BSG.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10 ?g/ml

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: MEM-M6/2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR