Anti-STAT1 Monoclonal Antibody [clone: PSM1]

Supplier: Abnova
MAB3641
10674-166EA 346 USD
10674-166
Anti-STAT1 Monoclonal Antibody [clone: PSM1]
Antibodies
Mouse monoclonal antibody raised against synthetic phosphopeptide of STAT1.

Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: PSM1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR