Anti-BUB3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12997
10703-534EA 527.69 USD
10703-534
Anti-BUB3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of BUB3.

Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: BUB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR