Anti-SOS1 Mouse Monoclonal Antibody [clone: SOS-01]

Supplier: Abnova
MAB3649
10739-044EA 406.15 USD
10739-044
Anti-SOS1 Mouse Monoclonal Antibody [clone: SOS-01]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of SOS1.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: SOS1
Clonality: Monoclonal
Clone: SOS-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR