Anti-HSV1 Mouse Monoclonal Antibody [clone: T96]

Supplier: Abnova
MAB3654
10718-402EA 329 USD
10718-402
Anti-HSV1 Mouse Monoclonal Antibody [clone: T96]
Antibodies
Mouse monoclonal antibody raised against recombinant herpes simplex virus type 1 (gC).

Type: Primary
Antigen: HSV1
Clonality: Monoclonal
Clone: T96
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR