Anti-Tubulin-beta Mouse Monoclonal Antibody [clone: TU-06]

Supplier: Abnova
MAB3656
10743-372EA 406.15 USD
10743-372
Anti-Tubulin-beta Mouse Monoclonal Antibody [clone: TU-06]
Antibodies
Mouse monoclonal antibody raised against native Tubulin-beta.

Recommended Dilutions: Immunocytochemistry (2 ?g/ml; Western Blot: 1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5 ?g/ml

Type: Primary
Antigen: Tubulin-beta
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: TU-06
Epitope:
Host: IgM
Isotype: Mouse
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR