Anti-Tubulin-alpha/beta dimer Mouse Monoclonal Antibody [clone: TU-08]

Supplier: Abnova
MAB3657
10743-366EA 406.15 USD
10743-366
Anti-Tubulin-alpha/beta dimer Mouse Monoclonal Antibody [clone: TU-08]
Antibodies
Mouse monoclonal antibody raised against native Tubulin.

Recommended Dilutions: Western Blot: 5 ?g/ml

Type: Primary
Antigen: Tubulin-alpha/beta dimer
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: TU-08
Epitope:
Host: IgM
Isotype: Mouse
Reactivity: Human, Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR