Anti-Tubulin-beta Mouse Monoclonal Antibody [clone: TU-13]

Supplier: Abnova
MAB3659
10743-374EA 329 USD
10743-374
Anti-Tubulin-beta Mouse Monoclonal Antibody [clone: TU-13]
Antibodies
Mouse monoclonal antibody raised against native Tubulin-beta.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: Tubulin-beta
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: TU-13
Epitope:
Host: IgM
Isotype: Mouse
Reactivity: Human, Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR