Anti-ZAP70 Mouse Monoclonal Antibody [clone: ZAP-03]

Supplier: Abnova
MAB3664
10745-018EA 406.15 USD
10745-018
Anti-ZAP70 Mouse Monoclonal Antibody [clone: ZAP-03]
Antibodies
Mouse monoclonal antibody raised against partial recombinant ZAP70.

Recommended Dilutions: Flow Cytometry: 2-5 ?g/ml; Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: ZAP-03
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR