Anti-POMC Mouse Monoclonal Antibody [clone: 180/A2 LF3]

Supplier: Abnova
MAB3665
10628-532EA 1178.46 USD
10628-532
Anti-POMC Mouse Monoclonal Antibody [clone: 180/A2 LF3]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of POMC.

Type: Primary
Antigen: POMC
Clonality: Monoclonal
Clone: 180/A2 LF3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR