Anti-CAMP Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13021
10704-832EA 527.69 USD
10704-832
Anti-CAMP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CAMP.

  • Recommended Dilutions: Western Blot: 1 μg/ml

Type: Primary
Antigen: CAMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR