Anti-AFP Mouse Monoclonal Antibody [clone: 2/23H5]

Supplier: Abnova
MAB3669
10626-516EA 530.63 USD
10626-516
Anti-AFP Mouse Monoclonal Antibody [clone: 2/23H5]
Antibodies
Mouse monoclonal antibody raised against recombinant AFP.

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: 2/23H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR