Anti-CD19 Mouse Monoclonal Antibody (Biotin) [clone: LT19]

Supplier: Abnova
MAB4370
10707-112EA 329 USD
10707-112
Anti-CD19 Mouse Monoclonal Antibody (Biotin) [clone: LT19]
Antibodies
Mouse monoclonal antibody raised against native CD19.

Recommended Dilutions: Flow Cytometry: 1:200

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: LT19
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR