Anti-PRKCD Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13641
10730-162EA 329 USD
10730-162
Anti-PRKCD Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Prkcd.

Recommended Dilutions: Western Blot: 1:250; Immunocytochemistry: 1:250; Immunohistochemistry: 1:250

Type: Primary
Antigen: PRKCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR