Anti-MS4A1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: LT20]

Supplier: Abnova
MAB4385
10725-948EA 329 USD
10725-948
Anti-MS4A1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: LT20]
Antibodies
Mouse monoclonal antibody raised against native MS4A1.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: LT20
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR