Anti-PPP2R2C Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13658
10733-736EA 638.45 USD
10733-736
Anti-PPP2R2C Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PPP2R2C.

Recommended Dilutions: Western Blot: 5-10 ?g/ml

Type: Primary
Antigen: PPP2R2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR