Anti-IL2RA Mouse Monoclonal Antibody (Biotin) [clone: MEM-140]

Supplier: Abnova
MAB4396
10721-302EA 329 USD
10721-302
Anti-IL2RA Mouse Monoclonal Antibody (Biotin) [clone: MEM-140]
Antibodies
Mouse monoclonal antibody raised against native IL2RA.

Recommended Dilutions: Flow Cytometry: 1:200

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: MEM-140
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR