Anti-CD27 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LT27]

Supplier: Abnova
MAB4397
10706-300EA 329 USD
10706-300
Anti-CD27 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LT27]
Antibodies
Mouse monoclonal antibody raised against native CD27.

Recommended Dilutions: Flow Cytometry: 10 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: LT27
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR