Anti-CYB5 Goat Polyclonal Antibody

Supplier: Abnova
PAB13663
10709-676EA 426.15 USD
10709-676
Anti-CYB5 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against CYB5.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CYB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR