Anti-CD33 Mouse Monoclonal Antibody (Biotin) [clone: HIM3-4]

Supplier: Abnova
MAB4406
10706-376EA 329 USD
10706-376
Anti-CD33 Mouse Monoclonal Antibody (Biotin) [clone: HIM3-4]
Antibodies
Mouse monoclonal antibody raised against native CD33.

Recommended Dilutions: Flow Cytometry: 1:1000

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: HIM3-4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR