Anti-DLX5 Goat Polyclonal Antibody

Supplier: Abnova
PAB13670
10710-916EA 428 USD
10710-916
Anti-DLX5 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of DLX5.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: DLX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR