Anti-GFP Mouse Monoclonal Antibody

Supplier: Abnova
MAB2003
10627-454EA 1170 USD
10627-454
Anti-GFP Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody raised against full length recombinant GFP.

Recommended Dilutions: ELISA: 1:200000; Western Blot: 1:3000-1:30000; Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: GFP
Clonality: Unconjugated
Clone: Monoclonal
Conjugation:
Epitope:
Host: IgG1 gamma
Isotype: Mouse
Reactivity: Jellyfishes
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR